Microcentrifuges, Pico™ and Fresco™
Supplier: Thermo Scientific
75002410 75002415 75002425 75002475 75002476 75002491 75002423 75002414 75002553
521-1095EA 2560 EUR
521-1095 521-1096 521-1098 521-1099 521-1100 521-1101 HERA75002423 HERA75002414 HERA75002553
Microcentrifuges, Pico™ and Fresco™
Centrifuges Microcentrifuges
Pico™ (ventilated) and Fresco™ (refrigerated) centrifuges are available in two speed options. Pico™ and Fresco™ 17 are perfect for routine laboratory work, while the Pico™ and Fresco™ 21 are ideal for high-end research applications.
  • The 24-place rotor can be used for all 1,5 to 2,0 ml tubes, including mini-preps and spin columns
  • Composite rotor technology, for greater chemical resistance and faster acceleration/deceleration compared to traditional aluminium
  • Unique ClickSeal® biocontainment rotor lid 'clicks' in place, providing assurance that samples are