HAEMAFlex™ 12/16 Swinging Bucket Rotors
Supplier: Thermo Scientific
75003862 75003882
HERA75003862EA 15190 EUR
HERA75003862 HERA75003882
HAEMAFlex™ 12/16 Swinging Bucket Rotors
Centrifuges Rotors
For high-throughput blood processing needs, the Thermo Scientific™ HAEMAFlex™ 12 swinging bucket rotor provides 12× 500 ml capacity or upgrade your centrifuge capacity to 16× 500 ml blood bags with the Thermo Scientific™ HAEMAFlex™ 16 rotor and the same bucket selections.
  • Stainless steel rotor body, aluminum windshield and bucket, polypropylene bottle
The rotors HAEMFlex 12 and 16 can be equipped with adapters to spin the most common blood collection tubes, conical tubes and plates, using oval buckets
These windshielded rotors feature ThermoMore Product Information