High-Performance Pharmacy Refrigerator, TSX
Supplier: Thermo Scientific
TSX2305PV TSX1205PV TSX3005PV TSX5005PV
HERATSX2305PVEA 8640 EUR
HERATSX2305PV HERATSX1205PV HERATSX3005PV HERATSX5005PV
High-Performance Pharmacy Refrigerator, TSX
Refrigerators
Thermo Scientific™ TSX Series high-performance pharmacy refrigerators are designed with features that support sample protection and sustainability objectives for the storage of pharmaceuticals, vaccines, chemotherapy and other medical and pharmacy-grade storage requiring 2 to 8 °C. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offeringMore Product Information