Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Supplier: HIRSCHMANN
344016060 344017060 344017560 344036060 344037060 344037560
612-9831EA 239 EUR
612-9831 HIRS344017060 HIRS344017560 HIRS344036060 HIRS344037060 HIRS344037560
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Burettes
Automatic burette, Pellet, class AS, DURAN® with blue graduation.
  • Lateral stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
  • Certified conformity, with dated batch identification
Certifikace: Compliance with DIN EN ISO 385.