Capillaries, Na-heparinised (coated), for blood gas analysis
Supplier: HIRSCHMANN
9221212 9221280 9222210 9222212 9222215 9222275 9221215 9221210 9221275
HIRS9221212EA 158 EUR
HIRS9221212 HIRS9221280 HIRS9222210 HIRS9222212 HIRS9222215 HIRS9222275 HIRS9221215 HIRS9221210 HIRS9221275
Capillaries, Na-heparinised (coated), for blood gas analysis
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Borosilicate glass 3.3.
  • Na-heparinized (coated)
For blood gas analysis.