Baths and circulators, with KISS® controller
Supplier: HUBER
2037.0040.98 2037.0052.98 2004.0009.98 2049.0003.98 2051.0003.98 2052.0003.98 2053.0003.98 2056.0004.98 2057.0004.98 2050.0003.98 2060.0004.98 2059.0004.98 2058.0004.98
HUBE2037.0040.98EA 1690 EUR
HUBE2037.0040.98 HUBE2037.0052.98 HUBE2004.0009.98 HUBE2049.0003.98 HUBE2051.0003.98 HUBE2052.0003.98 HUBE2053.0003.98 HUBE2056.0004.98 HUBE2057.0004.98 HUBE2050.0003.98 HUBE2060.0004.98 HUBE2059.0004.98 HUBE2058.0004.98
Baths and circulators, with KISS® controller
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
Heating circulation bath thermostats with either polycarbonate or stainless steel baths, which are simple to use and are suitable for a variety of applications such as: Sample temperature control, analysis, materials testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for Safety Class III/FL (DIN 12876) for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of externally attached applicationsMore Product Information