Baths and circulators, stainless steel
Supplier: HUBER
2038.0021.99 2057.0001.01 2056.0001.01 2058.0001.01 2059.0001.01 2060.0001.01
461-0557EA 0 EUR
461-0557 HUBE2057.0001.01 HUBE2056.0001.01 HUBE2058.0001.01 HUBE2059.0001.01 HUBE2060.0001.01
Baths and circulators, stainless steel
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
Circulating bath thermostats with baths made from stainless steel are simple to use and suitable for many applications such as: sample temperature control, analysis, material testing or checking food. The units offer high temperature stability and are fitted with over-temperature and low level protection suitable for use with flammable fluids. A powerful pressure/suction pump ensures optimum circulation and temperature homogenity in the bath and also allows temperature control of external applications using a pump adapter (accessory). There is a range of optional accessories includingMore Product Information