Circulating cooling thermostats, Minichiller and Unichillers
Supplier: HUBER
3013.0001.01 3013.0004.01 3015.0003.01 3024.0027.01 3006.0063.99 3014.0054.01 3014.0052.01 3024.0053.01 3024.0054.01 3025.0056.01 3010.0101.01 3065.0004.99 3066.0010.99 3055.0002.01 3055.0001.01 3054.0005.01 3054.0001.01 3054.0006.01 3054.0004.01 3054.0002.01 3013.0062.01 3014.0061.01 3052.0014.01 3025.0057.01 3013.0064.01 3051.0025.01 3024.0057.01 3024.0058.01
461-6472EA 0 EUR
461-6472 461-0988 461-1083 461-1085 461-0958 HUBE3014.0054.01 HUBE3014.0052.01 HUBE3024.0053.01 HUBE3024.0054.01 HUBE3025.0056.01 HUBE3010.0101.01 HUBE3065.0004.99 HUBE3066.0010.99 HUBE3055.0002.01 HUBE3055.0001.01 HUBE3054.0005.01 HUBE3054.0001.01 HUBE3054.0006.01 HUBE3054.0004.01 HUBE3054.0002.01 HUBE3013.0062.01 HUBE3014.0061.01 HUBE3052.0014.01 HUBE3025.0057.01 HUBE3013.0064.01 HUBE3051.0025.01 HUBE3024.0057.01 HUBE3024.0058.01
Circulating cooling thermostats, Minichiller and Unichillers
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Many applications depend on a reliable source of cooling. Circulating chillers in the Unichiller® range offer an ideal solution for environmentally friendly and economical cooling in laboratory and industry. There are over 50 air- and water-cooled models to choose from, with cooling powers from 0,3 to 50 kW. Efficient energy management ensures low operating costs and reduced usage of valuable fresh water. Huber circulating chillers are a resource saving solution which give a quick return on investment.
  • Robust stainless construction
  • Reliable continuous operation with warning functions
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Electrical 230 V, 50 Hz 208 V, 60 Hz 400 V, 50 Hz 220 - 240 V, 50 - 60 Hz 208 - 240 V, 50 - 60 Hz 115 V, 60 Hz 460 V, 60 Hz