MultiStore Storage system, Latch No. 6
Supplier: hünersdorff GmbH
460301
HUEN460301EA 0 EUR
HUEN460301
MultiStore Storage system, Latch No. 6
Boxes Storage Boxes
MultiStore Storage system no. 6 is versatile and can be used in places such as laboratories, workshops, sales rooms, doctor's surgeries, chemists, hospitals and industrial settings.
  • Bolts made of high-impact polystyrene, easily stackable and suitable for stable wall mounting
  • Dust-free and visible storage of the contents
  • For high loads and long life span
Removable tilting drawers made of crystal-clear polystyrene with blue plastic handle and labels for perfect drawer labelling.