Standard plates and mats, 96-well, WebSeal™
Supplier: Thermo Fisher Scientific
60180-P100 60180-P103 60180-P104 60180-P105 60180-P130 60180-P133 60180-P135
HYPE60180-P100EA 82.9 EUR
HYPE60180-P100 HYPE60180-P103 HYPE60180-P104 HYPE60180-P105 HYPE60180-P130 HYPE60180-P133 HYPE60180-P135
Standard plates and mats, 96-well, WebSeal™
Microplates Solid Supported Liquid/Liquid Extraction Plates (SLE)
WebSeal™ Standard Plates are plastic, non-coated, nonsterile, and chromatography tested.
  • WebSeal™ plates are manufactured from a GC/MS tested polypropylene material
  • Microplates are chemically and thermally resistant and will tolerate temperatures from will tolerate temperatures from −80 to 121 °C
  • SBS and ANSI standard footprint design for broad instrument compatibility
  • U- and V-bottom wells for optimal sample recovery
  • Low binding PP for homogenous assays and storage
WebSeal™ Mats and Sealing Tapes are nonsterile. Mats are manufactured of EVA (ethylene-More Product Information