Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 97, powder_white

Supplier: MP Biomedicals

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

0210121725 0210121780 0210121783 0210121790 0210121791
ICNA0210121725EA 42.6 EUR
ICNA0210121725 ICNA0210121780 ICNA0210121783 ICNA0210121790 ICNA0210121791
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 97, powder_white
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

Uridine-5'-triphosphate trisodium salt is a nucleoside diphosphate.


Uridine-5'-triphosphate trisodium salt is a P2Y receptor agonist.


UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients[3]. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes.


Soluble in water (50 mg/ml-clear to slightly hazy solution).


Upozornení: Research Use Only (RUO).

Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
MDL Number: MFCD00044310
CAS Number: 19817-92-6

Order Now

Specification Test Results

Identity Test Passes
Purity ≥97%
FTIR Conforms to Standard
OD 250/260 0.74 - 0.79
OD 280/260 0.32 - 0.40

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR