1,1'-Azobis(N,N-dimethylformamide), crystals_yellow

Supplier: MP Biomedicals

Synonyms: N1,N1,N2,N2-Tetramethyldiazene-1,2-dicarboxamide, Diamide

02101527.5 0210152701 0210152705 02101527.1
ICNA02101527.5EA 122 EUR
ICNA02101527.5 ICNA0210152701 ICNA0210152705 ICNA02101527.1
1,1'-Azobis(N,N-dimethylformamide), crystals_yellow
1,1'-Azobis(N,N-dimethylformamide)

Diamide is used as a thiol oxidising agent. It has also been used to titrate protein glutathiolation to discriminate from other oxidative protein modifications. Diamide treatment increased protein glutathiolation in a concentration-dependent manner and had comparably little effect on protein-protein disulphide formation.


Soluble in water.


Upozornení: Research Use Only (RUO).

Formula: C₆H₁₂N₄O₂
MW: 172.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00008318
CAS Number: 10465-78-8

Order Now

Specification Test Results

Identity Test Passes
Melting Point 104 - 115 °C
UV/Vis: λmax (water) 295 - 300 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR