(±)-2-Pyrrolidinone-5-carboxylic acid, powder_white

Supplier: MP Biomedicals

Synonyms: 5-Oxopyrrolidine-2-carboxylic acid, DL-2-Pyrrolidone-5-carboxylic acid, DL-Pyroglutamic acid

0210278080 0210278025
ICNA0210278080EA 80.8 EUR
ICNA0210278080 ICNA0210278025
(±)-2-Pyrrolidinone-5-carboxylic acid, powder_white
(±)-2-Pyrrolidinone-5-carboxylic acid

Soluble in 0,1 N HCI (clear, colourless solution), cold water and hot water.


Soluble in 0,1 N HCI (clear, colourless solution), cold water and hot water.

Formula: C₅H₇NO₃
MW: 129.12 g/mol
MDL Number: MFCD00064322
CAS Number: 149-87-1
EINECS: 205-748-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR