L(-)-Glutathione (oxidised form), powder_crystalline

Supplier: MP Biomedicals

Synonyms: L(-)-Glutathione, oxidized, Bi(glutathion-S-yl), GSSG, Glutathiol, Oxidized Glutathione (GSSG), Glutathione oxidized, (2S,2'S)-5,5'-(((2R,2'R)-Disulphanediylbis(1-((carboxymethyl)amino)-1-oxopropane-3,2-diyl))bis(azanediyl))bis(2-amino-5-oxopentanoic acid)

0215119305 02151193.2 02151193.5 0215119301
ICNA0215119305EA 244 EUR
ICNA0215119305 ICNA02151193.2 ICNA02151193.5 ICNA0215119301
L(-)-Glutathione (oxidised form), powder_crystalline
L(-)-Glutathione (oxidized form)

Amino Acid Sequence:Glu-Cys-Gly, Glu-Cys-Gly [disulfide bridge: 2a-2b].


In vivo, Glutathione disulfide (GSSG) is reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase. The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress


Soluble in water (3,3%): clear, colorless solution.


Upozornení: Research Use Only (RUO).

Formula: C₂₀H₃₂N₆O₁₂S₂
MW: 612.64 g/mol
Melting Pt: 185
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8

Order Now

Specification Test Results

Identity Test Passes
pH (3.3% aq soln) Acidic to Litmus Paper
Solubility (3.3% aq soln) Clear, Colorless Solution
Optical Rotation (c=1, water) –85 ° to –110 °
FTIR Conforms to Standard
Moisture For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR