Lecithin Soy Bean 90, solid_waxy orange-yellow chunky, Water Dispersable Grade

Supplier: MP Biomedicals

0215153925 0215153980 0215153983 0215153990
ICNA0215153925EA 175 EUR
ICNA0215153925 ICNA0215153980 ICNA0215153983 ICNA0215153990
Lecithin Soy Bean 90, solid_waxy orange-yellow chunky, Water Dispersable Grade
Lecithin

Lecithin is used as an emulsifier and solubilizer for drugs in an aqueous solution, also as a vehicle for liposomes.


Upozornení: Research Use Only (RUO).

Storage Temperature: Freezer
MDL Number: MFCD00147406
CAS Number: 8002-43-5
Merck Index: 12,05452

Order Now

Specification Test Results

Identity Test Passes
Purity ≥90%
Source Soybeans


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR