β-Caryophyllene, Liquid

Supplier: MP Biomedicals

Synonyms: (1R,4E,9S)-4,11,11-trimethyl-8-methylene-bicyclo[7.2.0]undec-4-ene, (1R,9S,E)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene, (−)-β-Caryophyllene, (-)-trans-caryophyllene, NSC 11906

0215494425 0215494405
ICNA0215494425EA 614 EUR
ICNA0215494425 ICNA0215494405
β-Caryophyllene, Liquid
β-Caryophyllene

(-)-trans-Caryophyllene is a natural bicyclic sesquiterpene that is a constituent of many essential oils.

Formula: C₁₅H₂₄
MW: 204.36 g/mol
MDL Number: MFCD00075925
CAS Number: 87-44-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR