Isoamyl alcohol 99, Liquid_clear, colorless

Supplier: MP Biomedicals
Danger

Synonyms: Isopentyl alcohol, iso-Pentanol, 3-Methyl-1-butanol, 3-Methylbutanol

0215507880 0215507890 0215507891
ICNA0215507880EA 32.7 EUR
ICNA0215507880 ICNA0215507890 ICNA0215507891
Isoamyl alcohol 99, Liquid_clear, colorless
Isoamyl alcohol

Isoamy alcohol is routinely used in molecular biology, especially in the purification of DNA. It is widely used in conjunction with phenol and chloroform (25:24:1 phenol : chloroform : isoamyl alcohol) for the removal of proteins from the nucleic acid solutions by extraction.


  • Japan fire code: 4.2
  • One of several isomers of amyl alcohol


Isoamyl alcohol is added to the phenol solution to help inhibit RNase activity and to help prevent the solubilisation in the phenol phase of long RNA molecules with long poly(A) portions. It will also help in reducing the foaming during the extraction process.Isoamyl alcohol is also used in the extraction of ethidium bromide from DNA solutions and in the radiolabeling of RNA transcripts in nuclei that have been isolated from tissue. Isoamy alcohol has various miscellaneous uses which include HPLC analysis of various pharmaceuticals and metabolites, nalysis of oxidised and reduced pyridine nucleotides and adenylates in organic phenol extracts from mitochondria, isolation of mRNA from a thermophilic cyanobacterium, Solvent for fats, resins, alkaloids, Manufacturing isoamyl (amyl) compounds, isovaleric acid, mercury fulminate, pyroxylin, artificial silk, lacquers, smokeless powders, for dehydrating celloidin solutions, for determining fat in milk etc.


Miscible with alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, oils.

Formula: (CH₃)₂CHCH₂CH₂OH
MW: 88.15 g/mol
Boiling Pt: 130
Melting Pt: –179 °F (lit.)
Flash Pt: 42
MDL Number: MFCD00002934
CAS Number: 123-51-3
UN: 1105
ADR: 3 III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR