Longifolene, Liquid_clear, colorless

Supplier: MP Biomedicals
Warning

Synonyms: (1S,3aR,4S,8aS)-4,8,8-Trimethyl-9-methylenedecahydro-1,4-methanoazulene, (+)-Longifolene

0215526501 0215526505
ICNA0215526501EA 53.6 EUR
ICNA0215526501 ICNA0215526505
Longifolene, Liquid_clear, colorless
Longifolene

(+)-Longifolene is a building block for the synthesis of hydroazulenes. It is also used in the preparation of dilongifolylborane, an effective chiral hydroborating agent.


Soluble in benzene; insoluble in water.

Formula: C₁₅H₂₄
MW: 204.357 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00082306
CAS Number: 475-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR