Methyl triacontanate, white powder

Supplier: MP Biomedicals

Synonyms: Mellisic acid methyl ester, Triacontanoic acid methyl ester

0215695980
ICNA0215695980EA 371 EUR
ICNA0215695980
Methyl triacontanate, white powder
Methyl triacontanate

Soluble in chloroform (50 mg/ml - clear colorless solution).

Formula: C₃₁H₆₂O₂
MW: 466.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042897
CAS Number: 629-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR