Adenosine-5'-diphosphate monosodium salt Equine muscle 97, powder

Supplier: MP Biomedicals

Synonyms: Adenosine 5'-diphosphate sodium salt, xSodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate, ADP-Na, Sodium ((2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate

02190149.1 02190149.5 0219014901
ICNA02190149.1EA 30.6 EUR
ICNA02190149.1 ICNA02190149.5 ICNA0219014901
Adenosine-5'-diphosphate monosodium salt Equine muscle 97, powder
Adenosine-5'-diphosphate monosodium salt
Formula: C₁₀H₁₄N₅NaO₁₀P₂
MW: 449.184 g/mol
MDL Number: MFCD00065987
CAS Number: 20398-34-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR