Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 90, powder_white

Supplier: MP Biomedicals

Synonyms: Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt

0219123090 0219123091 0219123080 0219123083
ICNA0219123090EA 142 EUR
ICNA0219123090 ICNA0219123091 ICNA0219123080 ICNA0219123083
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) 90, powder_white
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)

Energy-rich precursor in the enzymatic biosynthesis of RNA. A potent vasodilator. Induces contractile responses in some tissues.


Soluble in water (50 mg/ml - clear colourless solution).

Formula: C₉H₁₂N₂Na₃O₁₅P₃
MW: 550.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00044310
CAS Number: 19817-92-6
EINECS: 243-347-5

Order Now

Specification Test Results

Identity Test Passes
Purity ≥90%
OD250/OD260 0,75 ± 0,05
OD280/OD260 0,40 ± 0,05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR