Dimethyl sulphoxide 99,9, Liquid_clear, colorless ACS

Supplier: MP Biomedicals

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

0219141880 0219141890 0219141891
ICNA0219141880EA 40.2 EUR
ICNA0219141880 ICNA0219141890 ICNA0219141891
Dimethyl sulphoxide 99,9, Liquid_clear, colorless ACS
Dimethyl sulphoxide

Dimethyl sulfoxide is a dipolar, aprotic solvent. Has been shown to accelerate strand renaturation (1-10% concentration) and is believed to give the nucleic acid thermal stability against depurination.


  • Auto Ignition: 573 °F (Lit.)
  • Vapor Density: 2.7 (vs air) (lit.)
  • Vapor Pressure: 0.55 hPa at 20 °C
  • Refractive Index: n20/D 1.479(lit.)
  • Dielectric Constant: 45 (Lit.)


Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals. A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers. Used in antifreeze or hydraulic fluids. As a cryopreservative for cell cultures.Used in the oxidation of thiols and disulfides to sulfonic acids.Used as a PCR cosolvent to help improve yields, especially in long PCR.


Soluble in water, methanol, acetone, ether, benzene, chloroform.
Room Temperature, desiccate, store under nitrogen

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (lit.)
Melting Pt: 16…19 °C (lit.)
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 87
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay ≥99.9%
Residue after Evaporation ≤0.01%
Titratable Acid ≤0.001 meq/g
Water ≤0.1%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR