D-(+)-Glucose, anhydrous, powder_white Cell Culture

Supplier: MP Biomedicals

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

02194672.1 0219467201 0219467205
ICNA02194672.1EA 46.1 EUR
ICNA02194672.1 ICNA0219467201 ICNA0219467205
D-(+)-Glucose, anhydrous, powder_white Cell Culture
D(+)-Glucose

Storage: Store at Room Temperature (15-30 °C).
D- (+)-Glucose is a monosaccharide. It is a mixture of α- and β-anomers, primarily the α-anomer.
D- (+)-Glucose, anhydrous is a main source of energy for living organisms. It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.


  • Anhydrous
  • Cell Culture Reagent Grade
  • Presentation: White Powder
  • Soluble in 20% aqueous clear colorless solution.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 150…170 °C (lit.)
Density: 1.544 g/mL
Storage Temperature: Ambient
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Optical Rotation (c=10, Water, NH4OH) +52.5 ± 5°
Moisture ≤0.5%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR