Clostridium histolyticum Collagenase, Type I, MP Biomedicals
Supplier: MP Biomedicals
02195109.1 02195109.3 02195109.5 0219510901
ICNA02195109.1EA 94.2 EUR
ICNA02195109.1 ICNA02195109.3 ICNA02195109.5 ICNA0219510901
Clostridium histolyticum Collagenase, Type I, MP Biomedicals
Enzymes
Collagenase is a protease which cleaves the triple-helical protein called collagen. It has extensive use in biological studies, where it is used to prepare isolated cell suspensions.
  • Enzyme Commission Number: E.C.-3.4.24.3
It dissolve at a concentration of 1 mg/ml in 0,05 M TES with 0,36 mM calcium chloride, pH 7,5.
Unit definition: one unit will liberate peptides from collagen equivalent in ninhydrin color to 1,0 μmole of L-leucine in 5 hr at pH 7,4 and 37 °C in presence of calcium ions. Contains clostripain, neutral protease and trypsin activities.