Guanosine 5'-triphosphate disodium salt 95, powder_white

Supplier: MP Biomedicals
Warning

Synonyms: ((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt

0219521310 0219521325 0219521380 0219521383 0219521390 0219521391
ICNA0219521310EA 37.8 EUR
ICNA0219521310 ICNA0219521325 ICNA0219521380 ICNA0219521383 ICNA0219521390 ICNA0219521391
Guanosine 5'-triphosphate disodium salt 95, powder_white
Guanosine 5'-triphosphate disodium salt

Guanosine-5'-triphosphate, a purine nucleotide is required as a coenzyme for protein biosynthesis in a cell-free system.


Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exhange factors (GEF).


GTP functions as a carrier of phosphates and pyrophosphates involved in channeling chemical energy into specific biosynthetic pathways. GTP activates the signal transducing G proteins which are involved in various cellular processes including proliferation, differentiation, and activation of several intracellular kinase cascades. Proliferation and apoptosis are regulated in part by the hydrolysis of GTP by small GTPases Ras and Rho. Another type of small GTPase, Rab, plays a role in the docking and fusion of vesicles and may also be involved in vesicle formation. In addition to its role in signal transduction, GTP also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.

Formula: C₁₀H₁₄N₅Na₂O₁₄P₃
MW: 567.147 g/mol
Melting Pt: 206…207 °C (lit.)
Storage Temperature: Freezer
MDL Number: MFCD03410297
CAS Number: 56001-37-7
EINECS: 259-940-7

Order Now

Specification Test Results

Identity Test Passes
Purity ≥95%
FTIR Conforms to Standard
UV/VIS: l max (water) 252 ± 3nm
OD250/OD260 1.20 ± 0.08
OD280/OD260 0.65 ± 0.08
OD290/OD260 0.23 ± 0.08


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR