Cleaning solution for washing machines, concentrate, ES 7X-O-Matic®, MP Biomedicals
Supplier: MP Biomedicals
097667593 097667594 097667595 097667598
ICNA097667593EA 119 EUR
ICNA097667593 ICNA097667594 ICNA097667595 ICNA097667598
Cleaning solution for washing machines, concentrate, ES 7X-O-Matic®, MP Biomedicals
Cleaning Agents Detergents and Surface Cleansers
The ES 7X-O-Matic® solution is a low foaming, phosphate-free, concentrated surfactant and is particularly suited for automatic washing machines or procedures where the solution is agitated during cleaning.
  • Mildly alkaline, phosphate-free, safe and effective
  • Dark yellow liquid, soluble in water at any concentration
  • Contains powerful sequestering reagents
  • Ideal for cell cultureware and general labware
  • Cleans both glass and plastic labware
Solution contains a powerful wetting agent which will clean microscopic crevices not usually accessible to ordinaryMore Product Information