Handheld digital microscopes, Dino-Lite Medical, TrichoScope
Supplier: Dino-Lite / IDCP B.V.
MEDL3H MEDL7HM
630-2608EA 329 EUR
630-2608 IDCPMEDL7HM
Handheld digital microscopes, Dino-Lite Medical, TrichoScope
Microscopes
These handheld microscopes are ideal tools for studying the scalp, hair shaft and the hair itself in detail (trichoscopy). Images made with the TrichoScope can help the specialists to recognise skin or hair problems and diseases.
  • Built-in polariser filter (some models only) minimises the glaring effect of the scalp/hair
  • High speed USB 2.0 connection
  • Powered through USB connection: Do not need batteries
  • Polariser models: Built-in 8 LEDs can be enabled or disabled as desired; an external light source is unnecessary
  • Robust composite/plastic or metal housing