Sieves for Pilotina Mills
Supplier: IKA
U106469 U106470 U106471
IKAAU106469EA 0 EUR
IKAAU106469 IKAAU106470 IKAAU106471
Sieves for Pilotina Mills
Sieves Sieves for Mills
Interchangeable sieves for insertion into the grinding heads ensure maximum particle size filtering.
All sieves are made of stainless steel 1.4301.