D-(+)-Raffinose pentahydrate ≥98.0%, Ultrapure

Supplier: VWR Chemicals

Synonyms: D-(+)-Raffinose pentahydrate, Gossypose pentahydrate, (3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-((((3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-3,4,5-triol pentahydrate, Melitriose pentahydrate, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose pentahydrate

J392-100GEA 350 EUR
J392-100G
D-(+)-Raffinose pentahydrate ≥98.0%, Ultrapure
D(+)-Raffinose pentahydrate
Formula: C₁₈H₃₂O₁₆·5H₂O
MW: 594.52 g/mol
Melting Pt: 80
Density: 1.516 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00071590
CAS Number: 17629-30-0
EINECS: 208-146-9
Merck Index: 12,08279

Order Now

Specification Test Results

Arsenic <0.00005%
Heavy Metals <0.001%
Iron <0.0005%
Purity (HPLC) 98.0%
Residue on Ignition 0.1%
Solubility (10%, water) PASS
Specific Rotation (10%, Water) +103 to +107 °
Water Content (KF) 13.0 – 17.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR