Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Bestatin hydrochloride, (S)-2-((2S,3R)-3-Amino-2-hydroxy-4-phenylbutanamido)-4-methylpentanoic acid hydrochloride, Bestatin HCl

J60384.X0 J60384.MB
J60384.X0EA 107 EUR
J60384.X0 J60384.MB
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride) ≥98%
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Purity: 98% CAS No.: 65391-42-6 Molecular Formula: C16H24N2O4.HCl Formula Weight: 344.87 Physical Form: Powder Appearance: White Melting Point: 216-218° Beilstein Reference: 4628066 MDL No.: MFCD00058004


Aminopeptidase B inhibitor. Also inhibits leucine aminopeptidase

Formula: C₁₆H₂₅ClN₂O₄
MW: 308.4 · 36.5 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00058004
CAS Number: 65391-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR