EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M aqueous solution pH 8.0

Supplier: Thermo Fisher Scientific

Synonyms: EGTA, Ethylenebis(oxyethylenenitrilo)tetra(acetic acid), Kestranal® FE PA, Idranal® VI, Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, Ethylenebis(oxyethylenenitrilo)tetraacetic acid, Ethylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid, 3,12-Bis(carboxymethyl)-6,9-dioxa-3,12-diazatetradecanedioic acid, Titriplex® VI

J60767.ADEA 112 EUR
J60767.AD J60767.AE
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M aqueous solution pH 8.0
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)

CAS No.: 67-42-5 Molecular Formula: C14H24N2O10 Formula Weight: 380.35 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00004291

Formula: C₁₄H₂₄N₂O₁₀
MW: 380.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004291
CAS Number: 67-42-5
Merck Index: 13,03560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR