Methylene blue trihydrate

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride trihydrate, C.I.52015, Methylthioninium chloride trihydrate, Basic blue 9, Swiss Blue, Methylene blue

J60823.14 J60823.22 J60823.30
J60823.14EA 42.8 EUR
J60823.14 J60823.22 J60823.30
Methylene blue trihydrate
Methylene blue trihydrate

CAS No.: 7220-79-3 Molecular Formula: C16H18ClN3S.3H2O Formula Weight: 373.90 (319.86anhy) Physical Form: Crystalline Powder Appearance: Green Odour: Odourless Beilstein Reference: 3599847 Merck Reference: 14,6060 MDL No.: MFCD00150008


Sensitive stain for RNA and ribonuclease

Formula: C₁₆H₁₈ClN₃S·3H₂O
MW: 373.9 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150008
CAS Number: 7220-79-3
EINECS: 200-515-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR