Denatonium benzoate

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate

J61048.03 J61048.06 J61048.14
J61048.03EA 45 EUR
J61048.03 J61048.06 J61048.14
Denatonium benzoate
Denatonium benzoate

CAS No.: 3734-33-6 Molecular Formula: C21H29N2O.C7H5O2 Formula Weight: 446.59 Physical Form: Powder Appearance: White Melting Point: 164-169° Beilstein Reference: 8179408 Merck Reference: 14,2891 MDL No.: MFCD00031578


The bitterest compound known

Formula: C₂₈H₃₄N₂O₃
MW: 446.59 g/mol
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00031578
CAS Number: 3734-33-6
EINECS: 223-095-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR