Carboxy-PTIO potassium salt ≥98%

Supplier: Thermo Fisher Scientific

J61316.MA J61316.MF
J61316.MAEA 129 EUR
J61316.MA J61316.MF
Carboxy-PTIO potassium salt ≥98%
Carboxy-PTIO potassium salt

Purity: 98+% CAS No.: 148819-94-7 Molecular Formula: C14H16KN2O4 Formula Weight: 315.38 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Blue Solubility: Soluble in water. Also soluble in ethanol, methanol, DMSO or DMF. MDL No.: MFCD00216153


Applications: Stable, water-soluble free radical that reacts with nitric oxide (NO) to inhibit NO synthase

Formula: C₁₄H₁₆KN₂O₄
MW: 315.4 g/mol
MDL Number: MFCD00216153
CAS Number: 148819-94-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR