5-Iodo-2'-deoxycytidine 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidin-2(1H)-one

J61356.03 J61356.04
J61356.03EA 175 EUR
J61356.03 J61356.04
5-Iodo-2'-deoxycytidine 99%
5-Iodo-2'-deoxycytidine

Purity: 99% CAS No.: 611-53-0 Molecular Formula: C9H12IN3O4 Formula Weight: 353.11 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Melting Point: 180-183° MDL No.: MFCD00038063

Formula: C₉H₁₂IN₃O₄
MW: 353.12 g/mol
Melting Pt: 180…183 °C
MDL Number: MFCD00038063
CAS Number: 611-53-0
EINECS: 210-269-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR