Buffers, TRIS, 0,5 M solutions

Supplier: Thermo Fisher Scientific

J62287.AP J62287.K2 J61141.AP J61141.K2 J61144.AP J61144.K2 J62131.AP J62131.K2 J63735.AP J63735.K2
J62287.APEA 66.5 EUR
J62287.AP J62287.K2 J61141.AP J61141.K2 J61144.AP J61144.K2 J62131.AP J62131.K2 J63735.AP J63735.K2
Buffers, TRIS, 0,5 M solutions
Buffers Biological Buffers

CAS No.: 77-86-1
MDL No.: MFCD00004679

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form Liquid
CAS No. 77-86-1
MDL No. MFCD00004679

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR