Buffers, MES, 0,5 M solutions

Supplier: Thermo Fisher Scientific

J60031.AE J62534.AE J62534.AK J62574.AE J62574.AK J62081.AE J62081.AK J63778.AE J63778.AK
J60031.AEEA 52.5 EUR
J60031.AE J62534.AE J62534.AK J62574.AE J62574.AK J62081.AE J62081.AK J63778.AE J63778.AK
Buffers, MES, 0,5 M solutions
Buffers Biological Buffers

CAS No.: 145224-94-8
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00283848

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form Liquid
CAS No. 145224-94-8
MDL No. MFCD00283848
Appearance Colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR