Silver 0,02 mg/ml sodium citrate_2mM_supplied, nanoparticles 50 nm (Abs. 425 nm)

Supplier: Thermo Fisher Scientific

J67065.ACEA 96.4 EUR
J67065.AC J67065.AE
Silver 0,02 mg/ml sodium citrate_2mM_supplied, nanoparticles 50 nm (Abs. 425 nm)
Silver

CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Yellow Odour: Odourless MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Storage Temperature: Refrigerator
MDL Number: MFCD00003397
CAS Number: 7440-22-4
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR