Lightweight bump caps, HardCap AeroLite®
Supplier: JSP
AAF000-001-100 AAG000-001-100 AAG000-002-100
JSPAAAF000-001-100EA 60.3 EUR
JSPAAAF000-001-100 JSPAAAG000-001-100 JSPAAAG000-002-100
Lightweight bump caps, HardCap AeroLite®
Head Protection Bump Caps
One of the lightest full shell bump caps available on the market.
  • Weighs less than 135 g
  • EPP impact liner
  • Tough HDPE shell
  • Sleek baseball cap design
  • Polygiene® antibacterial coating
  • Air flow technology
  • Peak options
Weighing less than 135 g without compromising safety, HardCap AeroLite® bump caps are made using a lightweight EPP (Expanded Polypropylene) impact liner combined with a tough HDPE shell, all wrapped in a lightweight baseball style outer cap. Large reflective panels feature on the side of the HardCap AeroLite®, adding product and userMore Product Information