Floor cleaner, Quick Stripper, Ultan®
Supplier: KAMMERER
0404741
KAMM0404741EA 0 EUR
KAMM0404741
Floor cleaner, Quick Stripper, Ultan®
Cleaning Agents Detergents and Surface Cleansers
High performance intensive cleaner, quickly and effectively removes polish residues and caked-on dirt.
  • Odour-free
  • Only a small amount required
  • Suitable for all alkali-resistant floor surfaces