Sample tubes, with screw caps
Supplier: KARTELL
1671
KART1671EA 286 EUR
KART1671
Sample tubes, with screw caps
Tubes Test Tubes
PFA, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
With screw cap.