Reaction vessels, cylindrical with thermal and vacuum jacket, DN15 flange and drain valve
Supplier: KGW
TRGN7577
KGWATRGN7577EA 0 EUR
KGWATRGN7577
Reaction vessels, cylindrical with thermal and vacuum jacket, DN15 flange and drain valve
Reaction Vessels Reaction Vessels, Cylindrical Shape
This reaction vessel is made of borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With thermal and vacuum jacket, DN15 flange and drain valve.