Aluminium foil discs
Supplier: Korff
60206 60216 60289 60210 60220 60214 60272 60313 60312 60222 60232 60236 60271 60204 60235
611-9001EA 76.8 EUR
611-9001 611-9004 KORF60289 291-0050 291-0052 291-0051 KORF60272 KORF60313 KORF60312 KORF60222 KORF60232 KORF60236 KORF60271 KORF60204 KORF60235
Aluminium foil discs
Aluminium Foils
Pure aluminium, 99,0 to 99,5%.
  • Economical large packs
  • Tissue paper is inserted between discs to facilitate handling
  • Simple storage and removal from dispenser
Can be used for a wide variety of laboratory tasks.