Aluminium foil, cuttings
Supplier: Korff
60608 60278 60609 60629 60603 60642 60634 60631 60279 60619 60646 60265 60605 60612 60639 60282 60610
291-0029EA 43.1 EUR
291-0029 291-0030 291-0031 KORF60629 KORF60603 KORF60642 KORF60634 KORF60631 KORF60279 KORF60619 KORF60646 KORF60265 KORF60605 KORF60612 KORF60639 KORF60282 KORF60610
Aluminium foil, cuttings
Aluminium Foils
Pure aluminium 99,0 to 99,5%, soft.
  • Oxygen- gas- and water vapour-tight
  • Grease, moisture and water resistant
  • Heat resistant, fire resistant, non combustible
Ideal for packaging, covering and insulating. Hygienic and physiologically harmless. Resistant to most chemicals. Packed with an intermediate layer of tissue paper.