Gentamicin sulphate solution 10 mg/ml

Supplier: BIOWEST

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

L0011-010 L0011-100
L0011-010EA 14 EUR
L0011-010 L0011-100
Gentamicin sulphate solution 10 mg/ml
Gentamicin sulphate

Gentamicin sulfate acts against gram-negative, gram-positive bacteria and mycoplasma.


  • Storage temperature of –20 °C
  • pH of 4,5±0,3
  • Shelf life of 24 months
  • Endotoxin: <10 EU/ml (<1 ng/ml)


Used in cell culture applications at 5 ml/l. This concentration is for tissue culture media containing serum, serum-free media generally require lower concentration.


Upozornení: This product is intended to be used in vitro, in laboratory only. Not recommended to be used in therapy, human or veterinary applications.

MDL Number: MFCD00270181
CAS Number: 1405-41-0
Merck Index: 13,04403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR