2-Fluoro-6-methylpyridine 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Fluoro-6-picoline

L01561.06 L01561.03
L01561.06EA 65 EUR
L01561.06 L01561.03
2-Fluoro-6-methylpyridine 98
2-Fluoro-6-methylpyridine

Purity: 98% CAS No.: 407-22-7 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 66°(150°F) Refractive Index: 1.4700 Boiling Point: 140-141° Beilstein Reference: 107084 MDL No.: MFCD00041226

Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 140…141 °C
Density: 1.077
Flash Pt: 66 °C (150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041226
CAS Number: 407-22-7
EINECS: 206-980-8
UN: 1993
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR