4-Bromo-3-nitroanisole 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1-Bromo-4-methoxy-2-nitrobenzene

L06944.06 L06944.14
L06944.06EA 36 EUR
L06944.06 L06944.14
4-Bromo-3-nitroanisole 96%
4-Bromo-3-nitroanisole

Purity: 96% CAS No.: 5344-78-5 Molecular Formula: C7H6BrNO3 Formula Weight: 232.03 Flash Point: 110°(230°F) Melting Point: 29-32° Boiling Point: 153-154°/13mm Beilstein Reference: 2446617 MDL No.: MFCD00051511

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Boiling Pt: 153…154 °C (13 mmHg)
Melting Pt: 29…32 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051511
CAS Number: 5344-78-5
EINECS: 226-290-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR