2-Amino-1,3-propanediol 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Serinol, 2-Amino-1,3-dihydroxy propane, 2-Aminopropane-1,3-diol

L08426.06 L08426.14
L08426.06EA 77.8 EUR
L08426.06 L08426.14
2-Amino-1,3-propanediol 98%
2-Amino-1,3-propanediol

Purity: 98% CAS No.: 534-03-2 Molecular Formula: C3H9NO2 Formula Weight: 91.11 Melting Point: 52-56° Boiling Point: 128°/0.8mm UN No.: 3259 MDL No.: MFCD00051487

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 128 °C (0.8 mmHg)
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00051487
CAS Number: 534-03-2
EINECS: 208-584-0
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR