5-Fluoro-2-toluoyl chloride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-Fluoro-2-methylbenzene-1-carbonyl chloride, 5-Fluoro-2-methylbenzoyl chloride

L13750.03 L13750.06
L13750.03EA 27.8 EUR
L13750.03 L13750.06
5-Fluoro-2-toluoyl chloride ≥98%
5-Fluoro-2-toluoyl chloride

Purity: 98% CAS No.: 21900-39-0 Molecular Formula: C8H6ClFO Formula Weight: 172.59 Refractive Index: 1.5320 Boiling Point: 108°/24mm UN No.: 3265 MDL No.: MFCD00236681

Formula: C₈H₆ClFO
MW: 172.59 g/mol
Boiling Pt: 108 °C (24 mmHg)
Density: 1.270
Storage Temperature: Ambient
MDL Number: MFCD00236681
CAS Number: 21900-39-0
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR