N-Acetyl-L-leucine ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: N-Ac-Leu-OH, Ac-Leu-OH

L13926.06 L13926.14
L13926.06EA 39.2 EUR
L13926.06 L13926.14
N-Acetyl-L-leucine ≥99%
N-Acetyl-L-leucine

Purity: 99% CAS No.: 1188-21-2 Molecular Formula: C8H15NO3 Formula Weight: 173.21 Melting Point: 177-179° Beilstein Reference: 1724849

Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00065131
CAS Number: 1188-21-2
EINECS: 214-706-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR